missing translation for 'onlineSavingsMsg'
Learn More

Notrax Female MD-X Ramp System™ for Cushion Ease™ and SafetyStance™ Series

MD-X Ramp System™ is an extra wide attachable beveled safety ramp system for customized mat installations to avoid tripping. Compatible with the Notrax™ Cushion Ease™ and SafetyStance™ custom mats.

Brand:  Notrax 552F0003BL

 View more versions of this product

Product Code. 17722208

  • 49.00 € / Each
Estimated Shipment: 04-12-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • The 552 MD Extended Safety Ramp System™ Nitrile heavy duty bevels are compatible with both the CushionEase™ and SafetyStance™ Custom range
  • These bevels are 15 cm wide and 91 cm long and have been designed to provide easier access on and off the mat for things such as carts or trolleys
  • Their 100% nitrile rubber compound also ensures that these bevels are resistant to most oils making them perfect for heavier duty applications
  • It is available in black and yellow, which comply with OSHA code 1910-144
Specifications

Specifications

Black
91 cm
Industrial and Commercial Matting
36 in.
Nitrile Rubber
15 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers