missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fenbendazole sulfone, Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
10 mg
Description
Ensure you have the information you need to quickly start your analysis, and pass relevant audits:
- High quality inorganic and organic reference materials produced according to ISO 9001
- Certificate of Analysis (CoA) in every delivery complete with expiry date and lot specific analytical results
- Free expert technical support

Specifications
Specifications
CAS | 54029-20-8 |
Packaging | Glass Bottle |
Quantity | 10 mg |
Molecular Formula | C15H13N3O4S |
MDL Number | MFCD00797921 |
For Use With (Application) | Anthelmintics and Flukicides |
Beilstein | 703211 |
Physical Form | Crystalline Powder |
Molecular Weight (g/mol) | 331.35 |
Grade | Analytical Standard |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fenbendazole sulfone, Honeywell Fluka™ >
Spot an opportunity for improvement?Share a Content Correction