missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Bovine Serum Albumin (BSA)

Stabilize enzymes, increase PCR yields, and prevent adhesion of enzymes with Thermo Scientific™ Bovine Serum Albumin.

Brand:  Thermo Scientific™ B14

21.73 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 10829410

  • 30.60 € / 5mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Bovine Serum Albumin (BSA) is ideal for stabilization of enzymes during storage and for enzymatic reactions where the absence of nucleases is essential. BSA increases PCR yields from low purity templates. It also prevents adhesion of enzymes to the reaction tubes and tip surfaces.

Highlights

  • Non-modified (non-acetylated)
  • V-fraction
TRUSTED_SUSTAINABILITY
Specifications

Specifications

20 mg/mL
5 mg
Traditional Cloning
Molecular Biology
Bovine Serum Albumin
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2760344 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB14
Lot Number2832817 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberB14
Lot Number2864904 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberB14
Lot Number2869929 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberB14
Lot Number2889889 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberB14
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Bovine Serum Albumin (BSA) > 20mg/mL, 5mg (0.25mL)

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.