missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastDigest PstI

The FastDigest PstI restriction enzyme recognizes CTGCA^G sites and cuts best at 37°C in 5-15 minutes using universal FastDigest Buffer (Isoschizomers: BspMAI).

Brand:  Thermo Scientific™ FD0614

53.46 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10800421

  • 74.25 € / 800 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• FastDigest PstI (800 μL)
• 10X FastDigest Buffer (2 x 1.0 mL)
• 10X FastDigest Green Buffer (2 x 1.0 mL)

Store at –20°C.

Pst I
No
No
Restriction Enzyme
FastDigest™
Not Dam Methylation-Sensitive, Not Dcm Methylation-Sensitive, Not CpG Methylation-Sensitive
10X FastDigest Buffer/FastDigest Green Buffer
37°C
800 μL (800 reactions)
Traditional Cloning
3' protruding ends
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2851615 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog NumberFD0614
Lot Number2835171 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFD0614
Lot Number2883247 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberFD0614
Lot Number2890004 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberFD0614
Lot Number2910912 Certificate TypeCertificate of Analysis Date27/01/2025 Catalog NumberFD0614
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ FastDigest PstI > PstI; 800uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.