missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ FastDigest XbaI

Perform single-, double- or multiple DNA digestion within 5-15 minutes with this advanced line of restriction enzymes, 100% active in one universal buffer.

Brand:  Thermo Scientific™ FD0684

40.86 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


 View more versions of this product

Product Code. 10151320

  • 56.75 € / 300 reactions

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description
Show More
Specifications

Specifications

• FastDigest XbaI (300 μL)
• 10X FastDigest Buffer (1.0 mL)
• 10X FastDigest Green Buffer (1.0 mL)

Store at –20°C.

Xba I
Yes
No
Restriction Enzyme
FastDigest™
65°C
Dam Methylation-Sensitive, Not CpG Methylation-Sensitive, Not Dcm Methylation-Sensitive
10X FastDigest Buffer/FastDigest Green Buffer
37°C
300 μL (300 reactions)
Traditional Cloning
5' protruding ends
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Manuals

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2818378 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberFD0684
Lot Number2858727 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberFD0684
Lot Number2890621 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberFD0684
Lot Number2933253 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberFD0684
Lot Number2974424 Certificate TypeCertificate of Analysis Date26/01/2025 Catalog NumberFD0684
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ FastDigest XbaI > XbaI; 300uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.