Learn More
Thermo Scientific™ GeneJET Plant RNA Purification Kit
GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.
Brand: Thermo Scientific™ K0801
246.00 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.
Description
GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.
The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.
Thermo Scientific GeneJET Plant RNA Purification Mini Kit is designed for rapid and efficient purification of high quality total RNA from wide variety of plant species and tissue types.
The kit utilizes silica-based membrane technology in the form of a convenient spin column, eliminating the need for expensive resins, toxic phenol-chloroform extractions, or time-consuming alcohol precipitation. The standard procedure takes less than 20 minutes following cell lysis. Purified high quality RNA can be used in a wide range of downstream applications, such as RT-PCR, RT-qPCR, Northern blotting and other RNA-based analysis. RNA yields vary between different species, tissues, and age of tissue sample. See Table 1 for typical total RNA yields from various sources.
Highlights
• Fast—procedure completed in 20 min following cell lysis
• Convinient—protocol does not include clogging-prone lysate filtration step
• High yield—up to 65 μg of high purity and high integrity RNA from 50 mg plant sample
• Compatible with wide variety of samples (leaves, roots, sprouts and other parts of various plants)
Applications
Fast extraction of high purity RNA suitable for all conventional molecular biology procedures, including:
• RT-PCR
• RT-qPCR
• Northern blotting
• Nuclease protection assay
Specifications
• 40 mL Plant RNA Lysis Solution; store at room temperature • 40 mL Wash Buffer WB 1 (concentrated); room temperature • 30 mL Wash Buffer 2 (concentrated); room temperature • 30 mL Water, Nuclease-free; room temperature • 50 GeneJET RNA Spin Columns with Collection Tubes; room temperature • 50 Collection Tubes (2 mL); room temperature • 50 Collection Tubes (1.5 mL); room temperature |
|
20 min. (After sample homogenization) | |
Plants | |
Not High-throughput Compatible (Manual) | |
50 Preps | |
Room Temperature | |
Up to 65 μg |
Silica Spin Column | |
50 μL | |
Total RNA | |
RT-PCR, qPCR, cDNA Library Construction, NGS, Microarray Analysis, Blot Hybridization, Northern Blotting, In Vitro Translation, Nuclease Protection Assays, Nucleic Acid Labeling | |
Mini | |
≤100 mg |
Safety and Handling
- GeneJET Plant RNA Purification Kit
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 1 C
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H314-Causes severe skin burns and eye damage.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : Guanidine hydrochloride, Thiocyanic acid
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.