Learn More
Thermo Scientific™ GeneJET Plasmid Midiprep Kit
Designed for large-scale isolation of high quality plasmid DNA from recombinant E.coli cultures.
Brand: Thermo Scientific™ K0482
Description
Thermo Scientific GeneJET Plasmid Midiprep Kit, sufficient for 25 preps, is designed for large scale isolation of high quality plasmid DNA from recombinant E. coli cultures. The kit utilizes silica-based membrane technology in the form of convenient spin column. Each prep recovers up to 200 μg of high copy number plasmid DNA. The purified plasmid DNA can be used in a wide variety of molecular biology procedures, such as restriction endonuclease digestion, PCR, cloning, transformation, automated sequencing, in vitro transcription, and transfection of robust cell lines.
Highlights
- Efficient—high yields up to 200 μg of high quality plasmid DNA from 50 mL of bacterial culture
- High concentration of DNA purified— >400 ng/μL
- Convenient—spin column format, centrifuge or vacuum protocols
- Fast—procedure takes 60 minutes, considerably faster than using gravity flow columns
Applications
Isolated plasmid DNA is suitable for all conventional molecular biology procedures, including:
- FastDigest restriction enzyme digestion
- Automated fluorescent sequencing
- PCR
- Cloning
- Transformation
- In vitro transcription
- Transfection of robust cell lines

Specifications
400 ng/μL | |
Spin Column | |
Midi | |
Real-Time Quantitative PCR (qPCR), Transfection, Cloning, Sequencing, In Vitro Transcription, Transformation, Nucleic Acid Labeling, PCR | |
100-200 μg (Medium-Scale) Plasmid DNA | |
Plasmid MidiPrep Kit | |
Plasmid DNA |
Spin Column | |
Silica Column | |
Bacteria | |
Not High-throughput Compatible (Manual) | |
GeneJET | |
100 Preps | |
60 min. |
Safety and Handling
- GeneJET Plasmid Midiprep Kit
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
Hazard Statement
- H302-Harmful if swallowed.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P264-Wash hands thoroughly after handling.
- P270-Do not eat, drink or smoke when using this product.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P330-Rinse mouth.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : acetic acid, tritonX-100, guanidine hydrochloride
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.