missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ GeneRuler 1 kb DNA Ladder
GREENER_CHOICE

Perform sizing and approximate quantification of a wide range of double-stranded DNA on agarose gels with this mixture of chromatography-purified individual DNA fragments.

Brand:  Thermo Scientific™ SM0311

119.70 EUR valid until 2025-12-31
Use promo code "24358" to get your promotional price.


 View more versions of this product

Product Code. 11823963

  • 132.00 € / Pack of 5

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

GeneRuler 1kb DNA Ladder, range 250 to 10,000bp can be labeled radioactively with T4 Polynucleotide Kinase (seeProtocol).

The ladder is supplied with 6X DNA Loading Dye, and it can be used in DNA sizing and approximate quanitification applications.

The ladder is a mixture of chromatography-purified individual DNA fragments. GeneRuler 1kb DNA Ladder can be labeled radioactively with T4 Polynucleotide Kinase. The ladder is supplied with 6X DNA Loading Dye.
  • Ideal for both DNA sizing and approximate quantification
  • Sharp bands
  • Bright reference bands (given in red)
  • Supplied with loading dye for sample DNA
TRUSTED_SUSTAINABILITY
Specifications

Specifications

6X
Xylene Cyanol FF, Bromophenol Blue, Orange G
No
Agarose Gels
6X DNA Loading Dye, 2 x 1mL
5 x 50 μg
Agarose
DNA Ladder
250 to 10000 bp
Sustainable packaging
GeneRuler
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2815475 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM0311
Lot Number2829623 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM0311
Lot Number2815504 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM0311
Lot Number2801627 Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberSM0311
Lot Number2845647 Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberSM0311
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ GeneRuler 1 kb DNA Ladder > 1kb DNA ladder; 250-10000bp; 0.5ug/uL; 5 x 50ug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.