missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Maxima Probe/ROX qPCR Master Mix (2X)

Optimize qPCR with probe chemistry using standard cycling protocols with these ready-to-use qPCR master mixes.

Brand:  Thermo Scientific™ K0232

945.39 EUR valid until 2025-12-31
Use promo code "24271" to get your promotional price.


 View more versions of this product

Product Code. 11813923

  • 1112.00 € / 1000 reactions

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Maxima Probe qPCR Master Mixes are ready-to-use solutions optimized for quantitative real-time PCR. The master mixes include Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. Only template and primers need to be added.

Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carry-over contamination control using uracil DNA glycosylase (UDG). The use of Maxima Probe qPCR Master Mixes in real-time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral, and cDNA templates.

Maxima Probe qPCR Master Mixes are compatible with most real-time thermal cyclers.

The master mix includes Maxima Hot Start Taq DNA Polymerase and dNTPs in an optimized PCR buffer. It is supplemented with ROX passive reference dye. Only template and primers need to be added.Maxima Hot Start Taq DNA Polymerase in combination with an optimized buffer ensures PCR specificity and sensitivity. dUTP is included in the mix for optional carryover contamination control using uracil DNA glycosylase (UDG). ROX serves as a passive reference dye. The use of Maxima Probe/ROX qPCR Master Mix (2X) in real time PCR ensures reproducible, sensitive and specific quantification of genomic, plasmid, viral and cDNA templates. The master mix can be used with most real-time thermal cyclers, including instruments from Applied Biosystems, Eppendorf, Corbett, Bio-Rad and Roche.
  • Sensitivity—detects low copy number targets
  • Specificity—Maxima Hot Start Taq DNA Polymerase and the optimized buffer eliminate non-specific amplification and formation of primer dimers
  • Wide linear range—accurate quantification across 9 orders of magnitude
  • Universal—can be used with sequence-specific probes on most real-time thermal cyclers
  • Reproducibility and convenience—ready-to-use 2X master mix minimizes pipetting error and reduces set-up time

Recommended for:

Real-time PCR using sequence-specific probes; Real-time RT-PCR using sequence-specific probes.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Contains:
• 10 x 1.25 mL Maxima Probe/ROX qPCR Master Mix (2X) containing Maxima Hot Start Taq DNA Polymerase and dNTPs (also dUTP) in an optimized PCR buffer with ROX passive reference dye; sufficient for 1000 x 25 μL reactions.
• 10 x 1.25 mL Nuclease-Free Water

Store at -20°C.
Tube
qPCR
Standard
Maxima
1,000 reactions
Gene Expression
1,000 Reactions
10 x 1.25 mL
Primer-probe
High
Taq DNA Polymerase
7000 System, 7300 System, 7700 System, 7900HT System, StepOnePlus™ System, StepOne™ System
qPCR Master Mix
1000 Reactions at 25 μL
2X
DNA, dsDNA
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.