Learn More
Thermo Scientific™ RevertAid H Minus Reverse Transcriptase (200 U/μL)
Thermo Scientific RevertAid H Minus Reverse Transcriptase is a recombinant M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity, but lacks RNase H activity due to a point mutation in the RNase H domain.
193.00 € - 740.00 €
Specifications
Content And Storage | • RevertAid H Minus Reverse Transcriptase (200 U/μL) |
---|---|
Format | Stand-alone Enzyme |
Reaction Speed | 60 min. |
Technique | Reverse Transcription |
Optimal Reaction Temperature | 42°C to 45°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10121360
|
Thermo Scientific™
EP0451 |
10,000 Units |
193.00 €
10000 units |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11541515
|
Thermo Scientific™
EP0452 |
5 x 10,000 Units |
740.00 €
Pack of 5 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Thermo Scientific RevertAid H Minus Reverse Transcriptase is a recombinant M-MuLV RT. The enzyme possesses an RNA-dependent and DNA-dependent polymerase activity, but lacks RNase H activity due to a point mutation in the RNase H domain. RevertAid H Minus Reverse Transcriptase does not degrade RNA in RNA-DNA hybrids during synthesis of the first strand cDNA, and therefore high yields of full-length cDNA from long templates are obtained. RevertAid H Minus Reverse Transcriptase maintains activity over a wide temperature range (42–50°C).
Features of RevertAid H Minus Reverse Transcriptase include:
• High yields of full-length first strand cDNA up to 13 kb
• Optimum activity at 42–45°C
• Active up to 55°C
• Incorporates modified nucleotides (e.g., Cy3-, Cy5-, rhodamine-, aminoallyl-, and fluorescein-labeled nucleotides)
Applications
• First strand cDNA synthesis for RT-PCR and real-time RT-PCR
• Reverse transcription at elevated temperatures to reduce effects of secondary structure
• Synthesis of cDNA for cloning and expression
• Generation of labeled cDNA probes for microarrays
• DNA labeling
• Analysis of RNA by primer extension
Specifications
• RevertAid H Minus Reverse Transcriptase (200 U/μL) |
|
60 min. | |
42°C to 45°C | |
Reduced | |
200 U/μL | |
Separate components | |
Up to 13 kb |
Stand-alone Enzyme | |
Reverse Transcription | |
RevertAid H Minus | |
Dry Ice | |
First-Strand cDNA | |
Reverse Transcription | |
RNA |
For Research Use Only. Not for use in diagnostic procedures.