Learn More
Thermo Scientific™ RNase A/T1 Mix
Get the RNA degradation activity of both RNase A and RNase T1 with Thermo Scientific™ RNase A/T1 Mix.
Brand: Thermo Scientific™ EN0551
82.36 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.
Description
Thermo Scientific RNase A/T1 Mix combines the RNA degradation activity of both RNase A and RNase T1. The RNase A specifically hydrolyzes RNA at C and U residues; RNase T1 specifically hydrolyzes RNA at G residues (see Figure 1 in Supporting Data [see Reference 1]).
Highlights
- Higher level of RNA degradation than with RNase A and RNase T1 separately
- Free of DNase activity - It is not necessary to heat the mix before use
Specifications
RNase | |
1 mL | |
RNase A/T1 Mix |
Product Suggestions
Customers who viewed this item also viewed
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.