missing translation for 'onlineSavingsMsg'
Learn More

MP Biomedicals™ Fetal Bovine Serum, CELLect™ Gold

Superior for hybridoma cloning

Brand:  MP Biomedicals™ 092916849

 View more versions of this product

Product Code. 11481465

  • 380.00 € / 100mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Collected from a USDA regulated and monitored abattoir by technicians who work “onsite”
  • Aseptically collected via cardiac puncture
  • Supplied sterile, with no detectable mycoplasma or viral infection
TRUSTED_SUSTAINABILITY
Specifications

Specifications

USDA
-20°C
Fetal Bovine Serum
3.1 to 4.4 g/dL
Sterile
Used for cell culture and insect cell culture applications
27 mg/dL
280 to 325 mOsm/kg
USDA Inspected
Pale to Golden Brown
Liquid
1 x 100 mL
Heat-inactivated
<1 ng/mL
Yes
No
6.7 to 7.8
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.