missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Fetal Bovine Serum, CELLect™ Gold
Superior for hybridoma cloning
260.00 € - 1045.00 €
Specifications
Certifications/Compliance | USDA |
---|---|
Content And Storage | -20°C |
Form | Liquid |
Product Type | Fetal Bovine Serum |
Concentration | 3.1 to 4.4 g/dL |
Product Code | Brand | Color | Quantity | Sterilization Method | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Quantity | Sterilization Method | Price | Quantity & Availability | |||
11481465
|
MP Biomedicals™
092916849 |
Pale to Golden Brown | 1 x 100 mL |
380.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11471465
|
MP Biomedicals™
092916749 |
Golden Brown | 1 x 100 mL | Filtration |
260.00 €
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11445035
|
MP Biomedicals™
092916754 |
Golden Brown | 1 x 500 mL | Filtration |
1045.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
11455035
|
MP Biomedicals™
092916854 |
Pale to Golden Brown | 1 x 500 mL |
887.00 €
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Collected from a USDA regulated and monitored abattoir by technicians who work “onsite”
- Aseptically collected via cardiac puncture
- Supplied sterile, with no detectable mycoplasma or viral infection
Specifications
USDA | |
Liquid | |
3.1 to 4.4 g/dL | |
<1 ng/mL | |
No | |
6.7 to 7.8 |
-20°C | |
Fetal Bovine Serum | |
Sterile | |
27 mg/dL | |
280 to 325 mOsm/kg | |
USDA Inspected |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.