missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, charcoal stripped, USDA-approved regions
GREENER_CHOICE

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays.

Brand:  Gibco™ 12676029

 View more versions of this product

Product Code. 10706143

  • 730.00 € / 500mL

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco™ fetal bovine sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera Category: Specialty
  • Serum Treatment: Stripped with charcoal-dextran
  • Recommended Storage: -5°C to -20°C

Order Info

Shipping Condition: Dry Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Cattle/Bovine
Liquid
Fetal Bovine Serum
Charcoal-stripped
Dry Ice
Fetal
United States or Mexico
Bottle
500 mL
Research Grade
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.