missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, Premium Plus

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Brand:  Gibco™ A5669401

92.39 EUR valid until 2025-12-31
Use promo code "24268" to get your promotional price.


 View more versions of this product

Product Code. 17974721

  • 97.25 € / 50mL

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

Gibco fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements.

Sera category: Premium Plus

  • Use for a broad range of cell types, especially sensitive cell lines
  • Our most characterized serum with the lowest endotoxin and hemoglobin levels
  • Endotoxin level: ≤5 EU/mL
  • Hemoglobin level: ≤20 mg/dL
  • Origin: Refer to CoA for specific origin (US, Australia, New Zealand)
  • US Department of Agriculture (USDA) safety tested for the absence of bluetongue and akabane from countries that are USDA-approved for importation into the US

Gibco Serum Delivers

  • ISO 13485-certified; processed in FDA-registered facilities
  • Triple filtered at 0.1 μm
  • Gibco bottle easier to use in hood, reduces risk of contamination and helps improve consistency
  • Peel-off sticker on label provides handy reference for lot number and expiration date recording in lab notebook
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at ≤-10°C.
Fetal
Frozen
Fetal Bovine Serum
Standard (Sterile-filtered)
Triple-filtered, 0.1 μm
Triple-filtered, 0.1μm
≤5 EU/mL
≤20 mg/dL
USDA Safety Testing Performed for Australian origins
Cattle/Bovine
Refer to CoA for specific origin
One Shot
50 mL
Sterile
Premium Plus
Up to 70 quality tests including EMA virus testing; USP/EP mycoplasma, endotoxin, performance; biochemical/hormonal profiling; Oritain fingerprinting
Less waste
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberU2742007RP Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA5669401,A5669402
Lot NumberN2728118P Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA5669401,A5669402
Lot NumberN2751776P Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA5669401,A5669402
Lot NumberU2761908P Certificate TypeCertificate of Analysis Date29/01/2025 Catalog NumberA5669401,A5669402
Lot NumberN2813206P Certificate TypeCertificate of Analysis Date28/01/2025 Catalog NumberA5669401,A5669402
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Gibco™ Fetal Bovine Serum, Premium Plus > 50 mL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.