Learn More
Gibco™ Fetal Bovine Serum, Tet system approved, US origin
Gibco Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems.
Brand: Gibco™ A4736201
856.90 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Additional Details : Weight : 0.75000kg
Description
Gibco Tet system-approved FBS is formulated for cell culture applications using tetracycline- and tetracycline derivative-regulated gene expression systems. Cell-based assays that detect the presence of tetracycline and tetracycline analogs are used to select FBS that delivers the highest range of induction.
- Designed to ensure the absence of detectable tetracycline and its derivatives that may interfere with accurate regulation of gene expression
- Functional testing provides confidence that unintended tetracyline gene expression will not result
- Sera Category: Specialty
Specifications
Premium | |
Fetal | |
Cattle/Bovine | |
Liquid | |
Tet-System Approved | |
500 mL | |
Triple-filtered, 0.1 μm | |
<5 EU/mL | |
<15 mg/dL | |
Frozen/Dry Ice |
Store at <-10°C. | |
Use for a broad range of cell types, especially sensitive cell lines | |
United States | |
Bottle | |
Fetal Bovine Serum | |
Sterile | |
Meets USP/EP guidelines with up to 91 harmonized quality specification tests | |
Sustainable packaging | |
Premium |