missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Fetal Bovine Serum, ultra-low IgG, US origin, One Shot™ format
GREENER_CHOICE

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays.

Brand:  Gibco™ A3381901

 View more versions of this product

Product Code. 15695629

This item is not returnable. View return policy

Description

Description

Gibco™ fetal bovine sera offer excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

  • Sera category: Specialty
  • Origin: United States
  • Endotoxin level: ≤50 EU/mL (levels routinely ≤10 EU/mL)
  • Hemoglobin level: ≤25 mg/dL

IgG levels are less than 5 μg/mL, and the BVD titer is low or not detectable.

This reagent is classified as a medical device in several countries. If it is not regulated as a medical device in specific region, it should be used for laboratory use only.

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Storage conditions: ≤-10°C
Shipping conditions: Frozen
Bovine
Liquid
Fetal Bovine Serum
Exosome-depleted
One Shot™
Fetal
United States
One Shot
50 mL
Sterile
Frozen
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.