Learn More
Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor
GREENER_CHOICE
Conduct high-volume microplate processing with this large volume swinging bucket rotor.
Brand:  Thermo Scientific™ 75003665
4270.29 EUR valid until 2025-03-31
Use promo code "24119" to get your promotional price.
 View more versions of this product
Product Code. 10004124
  • 5507.40 € / Each
Explore more special offers
Description
Description
Conduct high-volume microplate processing with the Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor.
Features:
Fiberlite rotors:
  • Lightweight carbon fiber rotors reduce run-times by achieving higher G-forces
  • Corrosion-and fatigue-resistant design ensures structural integrity
  • Rotors are backed by a 15-year warranty and are compatible with Thermo Scientific™ Nalgene™ bottles and tubes for complete centrifuge solutions
Recommended for:
Pelleting cells and cellular debris, protein precipitation, plasmid purification and collecting physiological fluids for diagnostic tests
Note:
†Performance may vary by centrifuge model.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Fiberlite H3-LV Large Volume Swinging Bucket Rotor for Thermo Scientific Sorvall Legend XT, Multifuge X3 Series
28 Standard or 8 Deepwell Plates
3,600 rpm
90°
Energy efficient, less waste
Swinging Bucket Rotor
1,840 x g
Thermo Scientific Sorvall Legend XT/XF, Multifuge X3/X3F and SL 40F Centrifuges
Yes
Videos
SDS
Documents Certificates
View accessory products
Special Offers
Special Offers
Product Content Correction
Product Title
Thermo Scientific™ Fiberlite™ H3-LV Large Volume Swinging Bucket Rotor > For Fiberlite H3-LV Swinging Bucket Rotor