missing translation for 'onlineSavingsMsg'
Learn More

ROBU VitraPOR™ Filter Crucible, 50 mL

Crucibles are marked with an individual number and allow a distinct sample.

Brand:  ROBU VitraPOR™ 20504

 View more versions of this product

Product Code. 16843016

  • 221.00 € / Pack of 10
Estimated Shipment: 02-10-2024
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • For analytical filtration in laboratory
  • Suitable glass adapters and rubber sleeves are available
  • Highly resistant to most chemicals and temperatures up to 500°C
Specifications

Specifications

Borosilicate Glass (3.3)
10/Pk.
50 mL
4
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers