Learn More
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments offer easier fit, reliabililty and leak-free connections.
Brand:  Thermo Scientific™ 290SA133
 View more versions of this product
Product Code. 10137524
  • 90.90 € / Pack of 2
Estimated Shipment: 21-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments offer easier fit, reliabililty and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
  • Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
  • No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
  • Easy to handle with the nut touchable even with a hot injector/detector
  • No tools required
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Split/Splitless Injector Base Seal
Injector Base Seal; 2/Pk.
TRACE 1300 Series GC and Agilent GC Instruments
2/Pk.