Learn More
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments offer easier fit, reliabililty and leak-free connections.
90.90 € - 384.28 €
Specifications
For Use With (Equipment) TRACE 1300 Series GC and Agilent GC Instruments
View More Specs
Products 3
Product Code Brand Type Price Quantity & Availability  
Product Code Brand Type Price Quantity & Availability  
10794827
View Documents
Thermo Scientific™
290SA131
Split/Splitless and MS Starter Kit
384.28 €
Each
Estimated Shipment: 13-02-2025
to see stock.
Add to basket
10197494
View Documents
Thermo Scientific™
290SA132
Split/Splitless and FID Starter Kit
363.00 €
Each
Estimated Shipment: 05-02-2025
to see stock.
Add to basket
10137524
View Documents
Thermo Scientific™
290SA133
Split/Splitless Injector Base Seal
90.90 €
Pack of 2
Estimated Shipment: 21-01-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Finger Tite Connectors for Agilent Instruments offer easier fit, reliabililty and leak-free connections.
Advantages over conventional graphite/polymer ferrules:
  • Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
  • No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
  • Easy to handle with the nut touchable even with a hot injector/detector
  • No tools required
Specifications
Specifications
TRACE 1300 Series GC and Agilent GC Instruments