Learn More
Thermo Scientific™ Finger Tite Ferrules
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Brand:  Thermo Scientific™ 290S1132
 View more versions of this product
Product Code. 10015964
  • 104.35 € / Pack of 10
Estimated Shipment: 16-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific Finger Tite ferrules are the next generation of the SilTite™ connection system.
Advantages over conventional graphite/polymer ferrules:
  • Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
  • No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
  • No tools required
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Ferrule
0.4 mm ferrule
0.4 mm
0.1 to 0.25 mm I.D. GC Columns