Learn More
Thermo Scientific™ Finger Tite Ferrules
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
104.35 € - 106.75 €
Specifications
Type Ferrule
View More Specs
Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10055914
View Documents
Thermo Scientific™
290S1131
0.5 mm
106.75 €
Pack of 10
Estimated Shipment: 24-01-2025
to see stock.
Add to basket
10015964
View Documents
Thermo Scientific™
290S1132
0.4 mm
104.35 €
Pack of 10
Estimated Shipment: 16-01-2025
to see stock.
Add to basket
Description
Description
Thermo Scientific™ Finger Tite Ferrules offer easier fit, reliabililty and leak-free connections.
Thermo Scientific Finger Tite ferrules are the next generation of the SilTite™ connection system.
Advantages over conventional graphite/polymer ferrules:
  • Perfect metal-to-metal seal eliminates contamination and gives lower air/water background
  • No need to retighten ferrules, as they expand and contract with the fitting over GC operating temperatures
  • No tools required
Specifications
Specifications
Ferrule