Learn More
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes
GREENER_CHOICE
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of these fully autoclavable pipettes.
353.50 € - 493.00 €
Specifications
Description Finnpipette F2
Autoclavable Autoclavable
Number of Channels 1
Product Line Finnpipette™ F2
Type Variable Volume Pipette
View More Specs
Products 13
Product Code Brand Volume (Metric) Compatible Tips Color Increments Price Quantity & Availability  
Product Code Brand Volume (Metric) Compatible Tips Color Increments Price Quantity & Availability  
11877341
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642010
0.2 to 2 μL Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 Pink 0.002μL
371.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11817351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642020
0.5 to 5 μL Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 Pink 0.01μL
393.00 €
Each
In Stock
to see stock.
Add to basket
11827351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642030
1 to 10 μL Finntip 10 Micro, Filter 10 Micro, Flex 10, Flex Filter 10 Pink 0.02μL
353.50 €
Each
In Stock
to see stock.
Edge
Add to basket
11837351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642040
1 to 10 μL Finntip 250, Filter 10, Flex 200, Flex Filter 30, 200 Ext, Wide 250 Yellow 0.02μL
370.00 €
Each
In Stock
to see stock.
Add to basket
11847351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642050
2 to 20 μL Finntip 50 Micro, Filter 50 Micro Turquoise 0.02μL
370.00 €
Each
In Stock
to see stock.
Add to basket
10413865
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642060
2 to 20 μL Finntip 250, Filter 20, Flex 200, Flex Filter 30, 200 Ext, Wide 250 Yellow 0.02μL
474.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11867351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642070
10 to 100 μL Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, Wide 250 Yellow 0.2μL
370.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11877351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642080
20 to 200 μL Finntip 250, Filter 200, Flex 200, Flex Filter 200, 200 Ext, Filter 200 Ext, Wide 250 Yellow 0.2μL
474.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11887351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642090
100 to 1000 μL Finntip 1000, Filter 1000, Flex Filter 1000, 1000 Ext, Filter 1000 Ext, Wide 1000 Blue 1μL
474.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11897351
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642100
0.5 to 5 mL Finntip 5mL, Filter 5mL Green 0.01mL
493.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11807361
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642110
1 to 10 mL Finntip 10mL, Filter 10mL, Flex 10mL, Flex Filter 10mL Ext Red 0.02mL
482.00 €
Each
In Stock
to see stock.
Edge
Add to basket
11729528
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642120
5 to 50 μL Finntip 50 Micro, Filter 50 Micro Turquoise 0.1μL
368.00 €
Each
In Stock
to see stock.
Add to basket
11739528
GREENER_CHOICE
View accessory products View Documents
Thermo Scientific™
4642130
5 to 50 μL Finntip 250, Filter 100, Flex 200, Flex Filter 100, 200 Ext, Filter 100 Ext, Wide 250 Yellow 0.1μL
370.00 €
Each
In Stock
to see stock.
Edge
Add to basket
Description
Description
Meet the challenges of highly demanding applications where robustness and durability are essential with the sleek, rugged design of Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes. Built with tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light, these variable volume pipettes are fully autoclavable without disassembling. They feature a wide selection of expressly designed tips for optimal performance, precision and accuracy.
For decades, Thermo Scientific Finnpipette pipetting systems have delivered unmatched productivity and ergonomics across thousands of labs and applications worldwide.
Finnpipette F2: Proven reliable for long-term intensive use
Rugged and Durable
Finnpipette F2 contains tough PVDF components that stand up to harsh chemicals and the damaging effects of UV light. Its rugged design withstands physical use without damage.
Fully Autoclavable
Decontamination of the Finnpipette F2 is simple as there is no need to disassemble the F2 for autoclaving, minimizing disruption and downtime.
Maintenance is Easy
Simply detach the tip cone for efficient daily maintenance or decontamination when not using an autoclave.
Super Blow-out
150% increase in air boost to ensure efficient delivery of micro-volumes and prevent capillary action in 50μL models and below.
Wide Selection of Finntip Pipette Tips
Finntips are designed and manufactured expressly for Finnpipette models, enabling optimal performance, precision and accuracy.
5 Year Warranty
With convenient online registration, and may vary by country.
Specifications
Specifications
Finnpipette F2
1
Variable Volume Pipette
Autoclavable
Finnpipette™ F2
Videos
SDS
Documents
Documents
Product Certifications
Product Content Correction
Product Title
Thermo Scientific™ Finnpipette™ F2 Variable Volume Pipettes