missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Bovine Serum Albumin, Heat Shock Treated
Suitable for immunological studies
2825.00 €
Specifications
For Use With (Application) | Immunological Applications |
---|---|
pH Range | 7 ±0.3 |
Source | Bovine |
Packaging | Poly Bottle |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11403833
|
Fisher BioReagents™
BP1600-1 |
1 kg | N/A | ||||||
Description
CAS: 9048-46-8
Specifications
Immunological Applications | |
7 ±0.3 | |
Bovine | |
Poly Bottle |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Bovine Serum Albumin, Heat Shock, Reagent Grade Powder, pH 7.0
Signal Word
- Warning
Hazard Category
- Specific target organ toxicity Category 3
Hazard Statement
- H335-May cause respiratory irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.