Learn More
Fisher BioReagents™ EZ-Run™ Rec Protein Ladder
Mixture of 14 highly purified recombinant proteins optimized for precise sizing of proteins by SDS-PAGE and Western blotting
Brand:  Fisher BioReagents™ BP3602-500
 View more versions of this product
Product Code. 11498503
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Ladder is ideal for precise molecular weight determination of polypeptides in denaturing PAGE and Western blots
  • Each protein contains an integral Strep-tag™ II sequence1 and may be detected on Western blots using Strep-Tactin™-AP conjugate2 or an antibody against Strep-tag II
  • 50kDa protein band has greater intensity than other bands to serve as a reference
  • Supplied in gel loading buffer
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Protein Molecular Weight Marker/Ladder
Supplied in gel-loading buffer
<51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
10 to 200 kd
100 Loadings
Liquid
Molecular weight determination of polypeptides analyzed in various Gel system
500 μL, 100 Loadings
−20°C