missing translation for 'onlineSavingsMsg'
Learn More

Fisher BioReagents™ EZ-Run™ Rec Protein Ladder

Mixture of 14 highly purified recombinant proteins optimized for precise sizing of proteins by SDS-PAGE and Western blotting

113.00 € - 223.00 €

Specifications

Product Type Protein Molecular Weight Marker/Ladder
Form Liquid
Formulation Supplied in gel-loading buffer
For Use With (Application) Molecular weight determination of polypeptides analyzed in various Gel system
Kit Contents <51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
View More Specs

Products 2
Product Code Brand Quantity Sufficient For Price Quantity & Availability  
Product Code Brand Quantity Sufficient For Price Quantity & Availability  
11498503
View Documents
Fisher BioReagents™
BP3602-500
500 μL, 100 Loadings 100 Loadings This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10336693
View Documents
Fisher BioReagents™
BP3602-1
2 x 500 μL, 200 Loadings 200 Loadings This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Ladder is ideal for precise molecular weight determination of polypeptides in denaturing PAGE and Western blots
  • Each protein contains an integral Strep-tag™ II sequence1 and may be detected on Western blots using Strep-Tactin™-AP conjugate2 or an antibody against Strep-tag II
  • 50kDa protein band has greater intensity than other bands to serve as a reference
  • Supplied in gel loading buffer

Specifications

Specifications

Protein Molecular Weight Marker/Ladder
Supplied in gel-loading buffer
<51% Glycerin, 2% Sodium Lauryl Sulfate, <1% Tris 9Hydroxymethyl) Aminomethane, <0.76% 2,3-Butanediol, 1,4-Dimercapto, <0.5% Sodium Chloride, <0.05% Ethylenediaminet
−20°C
Liquid
Molecular weight determination of polypeptides analyzed in various Gel system
10 to 200 kd
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.