missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)
52.50 € - 391.00 €
Specifications
Color | White |
---|---|
For Use With (Application) | Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases |
Content And Storage | -20°C |
Source | Tritirachium album |
Product Type | Proteinase K |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10172903
|
Fisher BioReagents™
BP1700-50 |
50 mg |
52.50 €
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10103533
|
Fisher BioReagents™
BP1700-100 |
100 mg |
91.55 €
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10407583
|
Fisher BioReagents™
BP1700-500 |
500 mg |
391.00 €
500mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Proteinase K (Tritirachium album/Molecular Biology)
- Not on TSCA inventory: for research and development use only; not for manufacturing or commercial purposes
Proteinase K is widely used in the isolation of DNA and RNA from cell and tissue preparations. It is a wide-spectrum protease, inactivating many enzymes, including endogenous nucleases.
Specifications
White | |
-20°C | |
Proteinase K |
Isolation of DNA and RNA from cell and tissue preparations, inactivating many Enzymes, including Endogenous Nucleases | |
Tritirachium album | |
Solid |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Proteinase K
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
Hazard Statement
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Fisher BioReagents™ Proteinase K (Tritirachium album/Molecular Biology)