Learn More
Fisherbrand™ Air-Powered Analog Overhead Stirrers
Variable speed and explosion-proof stirrers ideal for safely mixing all types of solvents, lacquers and volatile chemicals without the possiblity of spark ignition.
Brand:  Fisherbrand™ 37020-9015
 View more versions of this product
Product Code. 15122997
  • 1320.00 € / Each
Estimated Shipment: 13-02-2025
to see stock.
Explore more special offers
Add to basket
Includes:
  • A 9/16 in. dia. x 10 in. L support rod
  • Four ft. air hose with double fittings for connection to air supply
Description
Description
  • Needle valve allows variation in mixing speed
  • Special muffler ensures quieter operation
  • Reversible models have motors which can be set to rotate in either direction by connecting the air supply line to the right port for counter-clockwise rotation or the left port for clockwise rotation
  • Can be used with flammable chemicals for explosion proof mixing
Order Info
Shaft, propeller, stand and clamp available separately
Specifications
Specifications
1000 rpm
0.37 in.
0.5
10 L x 0.56 in. dia. support rod, 4 ft. air hose with double fittings for connection to air supply
294
6 lb.
20 L
0.95 cm
Safely mixing all types of solvents, lacquers and volatile chemicals
30 to 80 psi
10,000 mPas
2.7 kg