missing translation for 'onlineSavingsMsg'
Learn More

Fisher Scientific™ Bath Circulator Accessories: Plumbing Packages

Plumbing kits for use with bath circulators and chillers

Brand:  Fisher Scientific™ 1600146

 View more versions of this product

Product Code. 11697988

  • 305.52 € / Each
Estimated Shipment: 21-03-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

Plumbing kits for use with bath circulators and chillers

  • Plumbing kit
  • Material: Viton
TRUSTED_SUSTAINABILITY
Specifications

Specifications

Thermo Scientific circulating bath
Plumbing Kit, Uninsulated
Viton Tubing
-25°C to +100°C
Package includes (4) clamps and (2) 5 ft. Viton tubing (uninsulated), temperature range of -30°C to +200°C, 12mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.