missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Probes, General purpose

Probes for use with Fisherbrand Traceable Thermometer.

Brand:  Fisherbrand™ 4001

 View more versions of this product

Product Code. 11789695

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Replacement probes
  • Made from stainless-steel

  • Replacement probes
  • Made from stainless-steel
Specifications

Specifications

0.12 in.
5 ft.
1500 mm
General triple purpose probe
300°F
-40°F
-40°F to +300°F
Digital Thermometer
3 mm
216 mm
Stainless Steel
8.5 in.
150°C
-40°C
-40°C to +150°C
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers