Learn More
Fisherbrand™ Sound Enclosure for Model 50 and 120 Sonic Dismembrator
Sound Enclosure for Model 50/120 Sonic Dismembrator. Sonic Dismembrators are extremely loud devices that can cause discomfort to the user and anyone nearby. The Sound Enclosure reduces noise by approximately 20 dBa and is made to work with all accessories. In addition to reducing noise, the Sound Enclosure has an internal support rod and converter mounting system. Any Fisherbrand probe or horn will be held safely and securely inside the unit.
Brand:  Fisherbrand™ FB432A
Product Code. 12353745
  • 1150.00 € / Each
Estimated Shipment: 06-06-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
This Fisher Scientific™ Sound Enclosure is designed for use with Model 50 and Model 120 Sonic Dismembrators
  • Sound Reducing Enclosure including stand and clamp
  • For use with Model 50/120 Sonic Dismembrator
  • Two ports are located on either side of the enclosure for coolant tubing or a temperature monitoring probe
  • Interior walls are lined with water-resistant acoustical foam
  • Door has a window so experiments can be visually monitored
  • Dimension: 50.8 x 25.4 x 25.4cm (20 x 10 x 10 in.)
Specifications
Specifications
Sound Enclosure
20 in.
Model 50/120 sonic dismembrator
12 x 12 x 20 in.
Ideal for sound reduction
Internal support rod and converter mounting system
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Sound Enclosure for Model 50 and 120 Sonic Dismembrator > Sound Enclosure with Clamp; 20 x 10 x 10 in.