Learn More
Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask
Manufactured from borosilicate glass. Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask's wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues.
Brand:  Quickfit FR4LF
 View more versions of this product
Product Code. 15547890
Description
Description
Manufactured from borosilicate glass
  • With large diameter Pyrex Labware™ Quickfit™ flat-flange joint
  • For use with Pyrex Labware™ Quickfit™ flat-flange lids and retaining clips
  • Wide neck allows easy incorporation of stirrer paddles and easy removal of solid residues
TRUSTED_SUSTAINABILITY
Specifications
Specifications
4 L
For use with Quickfit™ flat-flange lids and retaining clips
Wide
Borosilicate Glass
225 mm
Round
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Product Information
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Pyrex Labware™ Quickfit™ Borosilicate Glass Wide Neck Reaction Flask > Capacity: 4000mL