Learn More
Fisherbrand™ Stand and Clamp for Model 50 and 120 Sonic Dismembrator
Stand and Converter clamp accessory for Model 50/120 Sonic Dismembrator. This accessory will properly support the converter and probe assembly during sonication.
Brand: Fisherbrand™ FB460
85.51 EUR valid until 2024-12-31
Use promo code "21658" to get your promotional price.
Description
This Fisher Scientific™ Stand is designed for use with Model 50 and Model 120 Sonic Dismembrators
- Laboratory Support Stand with Converter clamp
- For use with Model 50/120 Sonic Dismembrator
Specifications
Stand | |
61 cm | |
0.5 in. dia. support rod and converter clamp |
24 in. | |
Model 50 and 120 sonic dismembrator |