Learn More
Fisherbrand™ Traceable™ Dual-Display Light Meter
Large LCD shows separate readings for lux and foot-candles in 1.375 in. H digits, plus selected source (daylight, tungsten, fluorescent or mercury).
Brand:  Fisherbrand™ 90023-86
Product Code. 11784266
  • 305.00 € / Each
In Stock
to see stock.
Explore more special offers
Add to basket
Description
Description
Large LCD shows separate readings for lux and foot-candles in 1.375 in. H digits, plus selected source (daylight, tungsten, fluorescent or mercury).
Three Ranges
  • Measures lux from 0 to 1999, 2000 to 19,990 and 20,000 to 50,000 (with resolutions ranging from 1 to 100 lux)
  • Measures foot-candles from 0 to 199.9, 200 to 1999 and 2000 to 5000 (with resolutions ranging from 0.1 to 10 ft.-c)
  • Accuracy is ±4.5% of full scale reading
Features
  • Zero reference points for daylight, tungsten, fluorescent and mercury; can report percent differential from reference point
  • Photodiode and color-correction filter provide cosine- and color-corrected measurements
  • Push-button access to highest, lowest and average readings
  • HOLD function freezes display to capture readings
  • Optional Data Acquisition System (11-661-22) or Data Logger System (06-662-72) required to transfer readings to computer
Order Info
Dimensions: 7L x 3W x 1.5 in.H (17.8 x 7.6 x 3.8cm); Net weight: 9.5 oz. (269g); Easy-Use™ 115V adapter (11-661-23), Data Acquisition System (11-661-22) and Data Logger System (06-662-72) available as accessories
Certifications
With individually serial-numbered ISO 17025 certificate indicating instrument traceability to standards provided by NIST™.
Specifications
Specifications
Light Meter
±4.5% of full scale reading
3.2 cm
7 in.
Dual Display
0 to 1999, 2000 to 19,990 and 20,000 to 50,000
1 to 100 lux
7.62 cm
±4.5% of full scale reading
With individually serial-numbered ISO 17025 certificate indicating instrument traceability to standards provided by NIST™.
2 in. Diameter (5.1cm) photocell, 40 in. (1m) cable, 9V alkaline battery, RS-232-C serial output for connection to IBM™-compatible computer or datalogger.
17.78 cm
0 to 199.9, 200 to 1999 and 2000 to 5000
0.1 to 10 ft.-c
3 in.
LCD
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
Product Content Correction
Product Title
Fisherbrand™ Traceable™ Dual-Display Light Meter > Light Meter For PC