missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™
IUPAC formulation
Brand: Fisherbrand 98769-33
This item is not returnable.
View return policy
Description
IUPAC formulation
- Compatible with all makes and instruments and electrodes
- Accuracy at 25°C is ±0.010 pH
Specifications
pH Buffer Standards | |
10.012 | |
Buffer | |
Bottle | |
Certified |
Instruments and Electrodes | |
16 oz. | |
Accuracy at 25°C is ±0010pH | |
Traceable to NIST |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™ > pH 10.012 16 oz. bottle
Spot an opportunity for improvement?Share a Content Correction