missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™
IUPAC formulation
19.54 € - 42.30 €
Specifications
Chemical Name or Material | pH Buffer Standards |
---|---|
For Use With (Application) | Calibration |
For Use With (Equipment) | Instruments and Electrodes |
Physical Form | Liquid |
Solution Type | Buffer |
Product Code | Brand | Quantity | Packaging | pH | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | pH | Price | Quantity & Availability | |||||
11774256
|
Fisherbrand
98769-34 |
6 x 100 mL | 6 shots of 100mL | 4.005 |
38.60 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11744256
|
Fisherbrand
90012-05 |
16 oz. | Bottle | 4.005 |
42.30 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11784256
|
Fisherbrand
98769-35 |
6 x 100 mL | 6 shots of 100mL | 7.00 |
38.60 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11754256
|
Fisherbrand
98769-32 |
16 oz. | Bottle | 7.0 | N/A | ||||||
11794256
|
Fisherbrand
98769-36 |
6 x 100 mL | 6 shots of 100mL | 10.012 |
38.10 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11764256
|
Fisherbrand
98769-33 |
16 oz. | Bottle | 10.012 | N/A | ||||||
Description
IUPAC formulation
- Compatible with all makes and instruments and electrodes
- Accuracy at 25°C is ±0.010 pH
Specifications
pH Buffer Standards | |
Instruments and Electrodes | |
Buffer | |
Traceable to NIST |
Calibration | |
Liquid | |
Accuracy at 25°C is ±0010pH | |
Certified |
Safety and Handling

Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Traceable™ pH Buffer Standards, Certified Reference Material (CRM), Fisherbrand™