missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Traceable™ Waterproof/Steamproof Timer

Used in food/beverage labs, wet labs, washdown areas, and for aquaculture, high humidity environments, field work.

Brand:  Fisherbrand™ 93678-98

31.26 EUR valid until 2025-04-30
Use promo code "24138" to get your promotional price.


Product Code. 15359433

  • 39.60 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Fisher Scientific™ Traceable™ Waterproof/Steamproof Timer is used in food/beverage labs, wet labs, washdown areas, and for aquaculture, high humidity environments, field work.

  • Resolution: 1 sec.
Specifications

Specifications

Waterproof/Steamproof Timer
Audible
1 sec.
2.75 oz.
Stand, removable suction cup, battery, Traceable certificate
NIST
99 min. 59 sec.
0.0001
Digital
77.96 g
Uses in food/beverage labs, wet labs, washdown areas, aquaculture, high humidity environments, field work
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Traceable™ Waterproof/Steamproof Timer > Waterproof/Steamproof Timer

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.