missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vortex Mixer Accessories: Foam Insert Set

For use with Fisherbrand vortex mixers.

Brand:  Fisherbrand™ 80862994

 View more versions of this product

Product Code. 15706949

  • 30.05 € / Each
Estimated Shipment: 12-03-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

For use with Fisherbrand vortex mixers.

Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.

Foam Insert Variety Pack

  • Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm

Specifications

Specifications

9 to 13 mm, 14 to 19 mm, 20 to 29 mm
Foam
9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
Foam Insert Set
14 to 19 mm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.