missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vortex Mixer Accessories: Foam Insert Set

For use with Fisherbrand vortex mixers.

29.30 € - 30.05 €

Specifications

Product Type Foam Insert Set
Material Foam
View More Specs

Products 3
Product Code Brand Tube Size For Use With (Equipment) Price Quantity & Availability  
Product Code Brand Tube Size For Use With (Equipment) Price Quantity & Availability  
15796939
View Documents
Fisherbrand™
80862993
9 to 13 mm Digital and analog mini vortex mixer
29.30 €
Each
Estimated Shipment: 12-02-2025
to see stock.
Add to basket
 
15706949
View Documents
Fisherbrand™
80862994
9 to 13 mm, 14 to 19 mm, 20 to 29 mm 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
30.05 €
Each
In Stock
to see stock.
Edge
Add to basket
 
15716949
View Documents
Fisherbrand™
80862995
20 to 25 mm 20 to 25mm Tube of digital and analog mini vortex mixer
29.30 €
Each
Estimated Shipment: 05-03-2025
to see stock.
Add to basket
 
Description

Description

For use with Fisherbrand vortex mixers.

Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.

Foam Insert Variety Pack

  • Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm

Specifications

Specifications

Foam Insert Set
Foam
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.