missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Vortex Mixer Accessories: Foam Insert Set
Description
For use with Fisherbrand vortex mixers.
Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.
Foam Insert Variety Pack
- Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm
Specifications
20 to 25 mm | |
Foam Insert Set | |
Foam | |
20 to 25mm Tube of digital and analog mini vortex mixer |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction