missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Vortex Mixer Accessories: Foam Insert Set

For use with Fisherbrand vortex mixers.

Brand:  Fisherbrand™ 80862993

 View more versions of this product

Product Code. 15796939

  • 29.30 € / Each
Estimated Shipment: 09-01-2025
to see stock.
Explore more special offers
Add to basket

Description

Description

For use with Fisherbrand vortex mixers.

Also compatible with other manufacturers' models; contact your Fisher Scientific Customer Service Representative for details.

Foam Insert Variety Pack

  • Includes foam inserts for the following: 9 to 13mm tube holder, 14 to 19mm tube holder, 20 to 29mm

Specifications

Specifications

9 to 13 mm
Foam Insert Set
Foam
Digital and analog mini vortex mixer
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers