Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
Brand:  Fisherbrand™ MP-20A
 View more versions of this product
Product Code. 12484657
  • 41.00 € / Each
Estimated Shipment: 17-02-2025
to see stock.
Explore more special offers
Add to basket
Specifications
Owl P8 Series Electrophoresis System
10 x 10 cm
0.05 cm
5.9 in.
15 cm
3.9 in.
13 cm
0.16 cm
13 cm
Comb
20
13 μL
3.9 in.
10 cm
0.06 in.
10 cm
5.11 in.