missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Fisherbrand™ Comb for use with FB-VE10-1 Vertical Electrophoresis System
For use with Fisherbrand™ FB-VE10-1 Vertical Electrophoresis System.
41.00 € - 42.30 €
Specifications
For Use With (Equipment) | Owl P8 Series Electrophoresis System |
---|---|
Type | Comb |
Gel Dimensions | 10 x 10 cm |
Product Code | Brand | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Teeth Count | Thickness (Metric) Teeth | Price | Quantity & Availability | ||||
12404667
|
Fisherbrand™
MP-8A |
8 | 0.77 cm |
41.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
11983015
|
Fisherbrand™
MP-6D |
6 | 0.15 cm |
41.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||||
11973015
|
Fisherbrand™
MP-6C |
6 | 0.08 cm |
41.00 €
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||
11993015
|
Fisherbrand™
MP-8C |
8 | 0.08 cm |
41.00 €
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||
11983025
|
Fisherbrand™
MP-8D |
8 | 0.15 cm |
41.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||||
11933015
|
Fisherbrand™
MP-10C |
10 | 0.08 cm |
42.10 €
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||
12404657
|
Fisherbrand™
MP-10A |
10 | 0.05 cm |
41.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
12464657
|
Fisherbrand™
MP-12A |
12 | 0.05 cm |
41.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
11953015
|
Fisherbrand™
MP-12C |
12 | 0.08 cm |
42.30 €
Each |
Estimated Shipment: 24-03-2025
Log in to see stock. |
||||
12484657
|
Fisherbrand™
MP-20A |
20 | 0.05 cm |
41.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
12474657
|
Fisherbrand™
MP-15A |
15 | 0.05 cm |
41.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
11963015
|
Fisherbrand™
MP-12D |
12 | 0.15 cm |
41.00 €
Each |
Estimated Shipment: 25-02-2025
Log in to see stock. |
||||
12494657
|
Fisherbrand™
MP-6A |
6 | 0.05 cm |
41.00 €
Each |
Estimated Shipment: 14-02-2025
Log in to see stock. |
||||
11943015
|
Fisherbrand™
MP-10D |
10 | 0.15 cm |
41.00 €
Each |
Estimated Shipment: 31-03-2025
Log in to see stock. |
||||
Description
Specifications
Owl P8 Series Electrophoresis System | |
Comb | |
10 x 10 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title