Learn More
Fisherbrand™ 384-Well ShallowWell Polypropylene Microplates
GREENER_CHOICE
Small volume applications
Brand:  Fisherbrand™ 267463
 View more versions of this product
Product Code. 13575450
  • 839.00 € / Pack of 100
Estimated Shipment: 25-03-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
Feature a shared-wall design which yields increased well volume
  • Automation compatible, ANSI standard
  • Low-binding and chemically resistant surface
  • Natural plates for storage or colorimetric assays
  • White plates for luminescent assays, including SPA
  • Black plates for fluorescent assays
  • Round bottom wells for optimal sample recovery
  • Rounded square wells minimize wicking (capillary action)
  • Working volume range: 2 to 35μL/well
Order Info
Lids not included
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ANSI
16 x 24
Polypropylene
384
Round, Conical Bottom (V-well)
Standard
White
58 μL
Sterile
Storage or colorimetric assays