missing translation for 'onlineSavingsMsg'
Learn More

Bel-Art™ SP Scienceware™ All-Metal Quick-Disconnects

Inline hose connector with automatic shutoff on disconnect to prevent leakage

Brand:  Bel-Art™ F19724-0204

 View more versions of this product

Product Code. 10107641

  • 72.00 € / Each
Estimated Shipment: 04-04-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • Chrome-plated brass hookups Compatible with most tubing
  • Operates at temperatures to 82°C (180°F), pressures to 250psi (1.7MPa) and vacuum to 30 in.Hg

Order Info

Order male and female connectors separately so tubing of the same or different size can be connected.

Specifications

Specifications

Quick Disconnect, Male Coupling
6.4 mm
6.4mm
0.25 in.
Metal
250 psi
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.