missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Biohazard Warning Tape
GREENER_CHOICE

Ideal for taping down bench protectors to delineate hazardous areas.

Brand:  Fisherbrand™ 15965

Product Code. 15809817

  • 5.20 € / Each
In Stock
to see stock.
Edge
Explore more special offers
Add to basket

Description

Description

Ideal for taping down bench protectors to delineate hazardous areas.

  • Adhesive tape is printed with the word “BIOHAZARD” and the biohazard warning symbol
  • Prevents curling and helps prevent spills from seeping under protective matting
  • Can be cut into labels (250 labels per roll)
  • Red fluorescent color

Order Info

Roll size: 500L x 0.50 in. W (1270 x 1.3cm).

Specifications

Specifications

Self-adhesive
Tape
Ideal for taping down bench protectors to delineate hazardous areas
500 in.
0.5 in.
Red
Biohazard Warning Tape
Each
12.7 m
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Biohazard Warning Tape > 500L x 0.50 in. W (1270 x 1.3cm) in roll

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.