missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Class B Amber Glass Threaded Vials with Attached Caps

Type I, Class B amber borosilicate glass

130.00 € - 426.40 €

Specifications

Classification B
Material (Liner) Teflon-faced Rubber
Color Amber
Material Amber Glass (Class B)
For Use With (Application) For general purpose vials for light sensitive materials
View More Specs

Includes: Attached black phenolic screw-top closures with PTFE-faced white 14B rubber liners

Products 5
Product Code Brand Volume (Metric) Height (Metric) Diameter (Metric) Outer Volume (English) Price Quantity & Availability  
Product Code Brand Volume (Metric) Height (Metric) Diameter (Metric) Outer Volume (English) Price Quantity & Availability  
11573542
View Documents
Fisherbrand™
90G14-955-321
2 mL 38 mm 12 mm 0.5 dr.
362.00 €
Pack of 288
Estimated Shipment: 13-05-2025
to see stock.
Add to basket
 
11513552
View Documents
Fisherbrand™
90X14-955-322
40 mL 98 mm 28 mm 10.82 dr.
256.00 €
Pack of 72
In Stock
to see stock.
Edge
Add to basket
 
11583542
View Documents
Fisherbrand™
90J14-955-331
4 mL 48 mm 15 mm 1 dr.
352.56 €
Pack of 144
Estimated Shipment: 17-07-2025
to see stock.
Add to basket
 
11309493
View Documents
Fisherbrand™
90L14-955-332
8 mL 63 mm 17 mm 2.16 dr.
426.40 €
Pack of 144
In Stock
to see stock.
Edge
Add to basket
 
11503552
View Documents
Fisherbrand™
90X14-955-333
20 mL 60 mm 28 mm 5.4 dr.
130.00 €
Pack of 72
In Stock
to see stock.
Edge
Add to basket
 
Description

Description

Type I, Class B amber borosilicate glass

  • General-purpose vials for light-sensitive materials
  • Made from Type I, Class B Amber Borosilicate Glass
  • Autoclavable

Compliance

Meet ASTM™ Type I, Class B and USP Type I standards

Specifications

Specifications

B
Amber
For general purpose vials for light sensitive materials
Autoclavable
ASTM, USP Type I
Teflon-faced Rubber
Amber Glass (Class B)
Phenolic
Fitted Cap
Vial
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Class B Amber Glass Threaded Vials with Attached Caps

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.