missing translation for 'onlineSavingsMsg'
Learn More

Fisherbrand™ Cryogenic Vial Storage Boxes
GREENER_CHOICE

Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials

Brand:  Fisherbrand™ 300462-0909

23.30 EUR valid until 2025-12-31
Use promo code "24332" to get your promotional price.


 View more versions of this product

Product Code. 11788029

  • 40.75 € / Pack of 4
In Stock
to see stock.
Explore more special offers
Add to basket

Description

Description

Suitable for ultra-low temperature storage of both internally and externally threaded cryogenic vials

  • 81-Place white polycarbonate storage boxes
  • Use in mechanical freezers and liquid nitrogen Dewars
  • Cover is printed with a 9 x 9 numbered grid for inventory purposes
  • Stackable
  • Autoclavable
TRUSTED_SUSTAINABILITY
Specifications

Specifications

White
Square
For ultra-low temperature storage of both internally and externally threaded cyrogenic vials
9 x 9
5.25 in.
5.25 in.
2 in.
4.31 kg
Polycarbonate
Cryo Storage Box
1.2 to 2 mL Threaded Cryo Vials
5.25 x 5.25 x 2 in. (13.3 x 13.3 x 5 cm)
13.3 cm
13.3 cm
5 cm
Product Suggestions

Product Suggestions

Videos
SDS
Documents Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Fisherbrand™ Cryogenic Vial Storage Boxes > For 1.2 to 2.0mL vials

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.