missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hellma Cylindrical Cells
Fit standard cell holders
221.00 € - 365.53 €
Product Code | Brand | Path Length | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Path Length | Includes | Price | Quantity & Availability | |||||
12711705
|
Hellma
120-10-40 |
10 mm | Suprasil quartz window; One stopper |
221.00 €
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
|||||
10126690
|
Hellma
120-50-40 |
50 mm | Suprasil quartz window; Two stopper |
365.53 €
Each |
Estimated Shipment: 17-06-2025
Log in to see stock. |
|||||
Description
Cylindrical cells have a filler neck and tapered PTFE stopper (two necks and stoppers on 50 and 100mm lightpath cells).
- Uniform diameter of 22mm
- Available with Suprasil™ quartz or near-UV glass windows
Specifications
Suprasil |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Hellma Cylindrical Cells